Learn More
Eppendorf™ Adapter for Conical Tubes
Product Code. 10725642 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor FA-45-6-30
Rotor FA-45-6-30
Unit Size:
Pack of 2
Pack of 7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10725642 Rotor FA-45-6-30 Pack of 2
10616640 Rotor A-4-81 and Rotor FA-45-6-30 Pack of 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10725642 Supplier Eppendorf™ Supplier No. 5820717009
  • ONLINE EXCLUSIVE PRICE
    1215.00 SEK
    1010.00 SEK / Paket med 2
    Save 205.00 SEK
    16% Av
 Restricted Item
I lager
Designed for use with a variety of products to expand liquid handling capabilities in the lab
  • Available in packs of two or seven
  • For use with Eppendorf Rotors
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical
Capacity 15 mL
For Use With (Equipment) Rotor FA-45-6-30
Color Orange
No. per Pack Set of 2