Learn More
Hettich™ Adapter for Rotors no. 4266, 4570, 4795 and 5645
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5646
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12093924
  • 855.00 SEK / Each
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
  • Adapter for Rotors no. 4266, 4795, 5641 and 5645
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
7 x 15 mL
100 mm
7
Round Bottom
17 mm
Rotors no. 4266, 4570, 4795 and 5645