missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Adapter for Rotors no. 4266, 4570, 4795 and 5645

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 5641

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12083924

  • 1015.00 SEK / Each

Estimated Shipment: 08-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Adapter for Rotors no. 4266, 4795, 5641 and 5645
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications

Specifications

16 mm
10 mL Tubes
Rotors no. 4266, 4570, 4795 and 5645
Rotor Adapter
80 mm
Round Bottom
8
8
Videos
SDS
Documents
Certificates