Läs mer
Adenosine-13C5, TRC
Produktkod. 30011374
Change view
Kvantitet:
2.5 mg
25 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011374 25 mg -
30011120 2.5 mg 2.5mg
30009431 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA28040225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 159496-13-6
Molekylformel C513C5H13N5O4
Molekylvikt (g/mol) 272.2
Synonym 9-β-D-Ribofuranosyl-9H-purin-6-amine-13C5, 9-β-D-Ribofuranosyladenine-13C5, Adenine Riboside-13C5, Adenocard-13C5, Adenocor-13C5, Adenogesic-13C5, Adenoscan-13C5, Adrekar-13C5, Boniton-13C5, D-Adenosine-13C5, 1-(6-Amino-9H-purin-9-yl)-1-deoxy-β-D-ribofuranose-13C5, Myocol-13C5, NSC 7652-13C5;
IUPAC-namn (2R,3S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)CO)O)O
Specifikationer
Kemiskt namn eller material Adenosine-13C5
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6?,7+,10-/m1/s1/i1+1,4+1,6+1,7+1,10+1
Formel vikt 272.114
Procent renhet >95%