Learn More
Adenosine Deaminase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554912
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 88%.
Adenosine Deaminase Polyclonal antibody specifically detects Adenosine Deaminase in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Adenosine Deaminase | |
Polyclonal | |
Unconjugated | |
ADA | |
ada; ADA1; adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5; zgc:92028 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00813 | |
ADA | |
Recombinant protein corresponding to Human Adenosine Deaminase | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only