missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Adenovirus-9 E4 Orf1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41088B
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Adenovirus-9 E4 Orf1 Polyclonal antibody specifically detects Adenovirus-9 E4 Orf1 in Adenovirus, Virus samples. It is validated for ELISA
Specifikationer
| Adenovirus-9 E4 Orf1 | |
| Polyclonal | |
| ELISA | |
| 13.6kDa protein, 14.5kDa protein, Control protein orf2, Control protein orf3, Control protein orf4, E4, E4 13.5kDa protein, E4 13.6kDa protein, E4 14.4kDa protein, E4 14.5kDa protein, E4 control protein 34 kDa, E4 control protein orf2, E4 control protein orf3, E4 control protein orf4, HuAdVD_gp17, ORF 2, Orf 3, Orf 4, ORF2, ORF3, ORF3 protein, ORF4, Putative E4 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the Ad-9 E4 Orf1. The immunogen is located within the last 50 amino acids of Adenovirus-9 E4 Orf1. Amino Acid Squence: FNHTDRFLYVREGHPV | |
| 0.1 mL | |
| Cancer | |
| 6386282 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Adenovirus, Virus | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering