missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Adenovirus Type 2 and 5 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.
1150.00 SEK - 6605.00 SEK
Specifications
Antigen | Adenovirus Type 2 and 5 Ab-5 |
---|---|
Clone | M58 + M73 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12688826
|
Epredia™
MS-1069-PABX |
Concentrated | 200 μL |
6605.00 SEK
200µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12668826
|
Epredia™
MS-1069-P1 |
Concentrated | 500 μL |
4200.00 SEK
500µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12648826
|
Epredia™
MS-1069-P |
Concentrated | 1 mL |
6045.00 SEK
1mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12618836
|
Epredia™
MS-1069-R7 |
Prediluted | 7 mL |
2985.00 SEK
7mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12658826
|
Epredia™
MS-1069-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12678826
|
Epredia™
MS-1069-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Adenovirus Ab-5 detects adenovirus serogroups 2 and 5. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.Host Species: Mouse
Clone: M58 + M73
Isotype: IgG2a/κ + IgG2a/κ
Species Reactivity: Adenovirus.
Immunogen: Adenovirus
Positive Control: 293 cells or adenovirus infected tissues
Cellular Localization: Nuclear.
Ab-5 is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Adenovirus Type 2 and 5 Ab-5 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Adenovirus | |
IgG2a κ |
M58 + M73 | |
Monoclonal | |
Mouse | |
Infectious Disease | |
Adenovirus | |
Primary |