Learn More
3M™ Adflo™ Odor Filter Pad
3M™ Adflo™ Odor Filter Pad is a spare filter that should be replace any time you smell unpleasant odors.
Brand: 3M™ 7000044586
415.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Odor filter is not required if gas filter is used
Specifications
Black | |
Spare Filter Pad |