Learn More
Brady™ Adhesive Polyethylene Fire Extinguisher Pictograms
Product Code. 11760564 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
L x H: 200 x 250 mm
L x H: 315 x 315 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11760564 L x H: 200 x 250 mm Each
11730853 L x H: 315 x 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11760564 Supplier Brady™ Supplier No. 251127
  • ONLINE EXCLUSIVE PRICE
    64.55 SEK
    54.38 SEK / Paket med 1
    Save 10.17 SEK
    15% Av
Beräknad leverans: 10-12-2026