Läs mer
Adipic Acid-13C6, TRC
Produktkod. 30007855
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007855 10 mg
30006758 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA29159310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 942037-55-0
Molekylformel 13C6H10O4
Molekylvikt (g/mol) 152.1
Synonym 1,4-Butanedicarboxylic Acid-13C6, 1,6-Hexanedioic Acid-13C6, Acifloctin-13C6, Acinetten-13C6, Adilactetten-13C6, Adipinic Acid-13C6, Asapic-13C6, E 355-13C6, Inipol DS-13C6, NSC 7622-13C6, NSC 87836-13C6
IUPAC-namn (1,2,3,4,5,6-13C6)hexanedioic acid
LEDER O[13C]([13C]([H])([H])[13C]([H])([H])[13C]([H])([H])[13C]([H])([H])[13C](O)=O)=O
Specifikationer
Kemiskt namn eller material Adipic Acid-13C6
Kvantitet 10 mg
InChI formel InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1