Läs mer
Adipokinetic Hormone (Anax Imperator Mauricianus), TRC
Produktkod. 30312340
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30312340 100 mg
1 options
Produktkod. 30312340 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA291645100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Adipokinetic Hormone (Anax Imperator Mauricianus)
CAS 154512-22-8
Molekylformel C45H59N11O12
Kvantitet 100 mg
Synonym 5-oxo-L-prolyl-L-valyl-L-asparaginyl-L-phenylalanyl-L-seryl-L-prolyl-L-seryl-L-Tryptophanamide, Chromatophorotropin, red-pigment-concentrating (Pandalus borealis) 2-L-valine-7-L-serine-, 1686: PN: US20040102389 SEQID: 10049 claimed protein, Adipokinetic hormone (Anax imperator), Anaim-AKH, Ani-AKH
InChI formel InChI=1S/C45H59N11O12/c1-23(2)37(55-39(62)28-14-15-36(60)49-28)44(67)52-31(19-35(46)59)41(64)51-30(17-24-9-4-3-5-10-24)40(63)54-33(22-58)45(68)56-16-8-13-34(56)43(66)53-32(21-57)42(65)50-29(38(47)61)18-25-20-48-27-12-7-6-11-26(25)27/h3-7,9-12,20,23,28-34,37,48,57-58H,8,13-19,21-22H2,1-2H3,(H2,46,59)(H2,47,61)(H,49,60)(H,50,65)(H,51,64)(H,52,67)(H,53,66)(H,54,63)(H,55,62)/t28-,29-,30-,31+,32-,33-,34?,37+/m0/s1
LEDER CC(C)[C@@H](NC(=O)[C@@H]1CCC(=O)N1)C(=O)N[C@H](CC(=O)N)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](CO)C(=O)N3CCCC3C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N
IUPAC-namn (2R)-N-[(2S)-1-[[(2S)-1-[2-[[(2S)-1-[[(2S)-1-amino-3-(1H-indol-3-yl)-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]carbamoyl]pyrrolidin-1-yl]-3-hydroxy-1-oxopropan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]-2-[[(2R)-3-methyl-2-[[(2S)-5-oxopyrrolidine-2-carbonyl]amino]butanoyl]amino]butanediamide
Molekylvikt (g/mol) 946.02
Formel vikt 945.4345