missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Adiponectin Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16382145
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16382145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16382145 Leverantör Invitrogen™ Leverantörsnummer PA585206

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (100%). Positive Control: MCF-7, Mouse white adipose tissue, mouse muscle, rat muscle. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Adiponectin (also known as 30- kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR-. Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Adiponectin Receptor 1
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gen ADIPOR1
Genaccessionsnr. Q91VH1, Q96A54
Gene Alias 2810031L11Rik; ACDCR1; adiponectin receptor 1; adiponectin receptor protein 1; Adipor1; CGI45; CGI-45; PAQR1; Parq1; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member I; TESBP1A
Gensymboler ADIPOR1
Värd art Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human Adiponectin Receptor 1. The exact sequence is proprietary.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 289036, 51094, 72674
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.