missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADIPOQ Mouse anti-Human, Mouse, Clone: 5D8A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66239-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.Specifications
ADIPOQ | |
Monoclonal | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15848, Q60994 | |
ADIPOQ | |
ADIPOQ Fusion Protein Ag17383 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
5D8A7 | |
Unconjugated | |
ADIPOQ | |
ACDC, ACRP30, adiponectin, ADIPOQ, ADIPQTL1, ADPN, APM 1, APM1, GBP28, Gelatin binding protein | |
Mouse | |
Protein G | |
RUO | |
11450, 9370 | |
-20°C | |
Liquid |