Läs mer
Adrenalone Hydrochloride hydrate, TRC
Produktkod. 30136873
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136873 10 g
30007809 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136873 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA30510010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62-13-5
Molekylformel C9 H11 N O3 . Cl H
Molekylvikt (g/mol) 217.65
Synonym 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride, Adrenaline Tartrate Imp. C (EP) as Hydrochloride, Adrenalone Hydrochloride, Ethanone, 1-(3,4-dihydroxyphenyl)-2-(methylamino)-, hydrochloride (1:1), Acetophenone, 3',4'-dihydroxy-2-(methylamino)-, hydrochloride (7CI,8CI), Adrenalone hydrochloride (6CI), Ethanone, 1-(3,4-dihydroxyphenyl)-2-(methylamino)-, hydrochloride (9CI), Adrenone hydrochloride, Kephrine hydrochloride, Styphnonasal, α-Methylamino-3,4-dihydroxyacetophenone hydrochloride, α-Methylamino-3',4'-dihydroxyacetophenone hydrochloride
IUPAC-namn 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanone;hydrochloride
LEDER Cl.CNCC(=O)c1ccc(O)c(O)c1
Specifikationer
Kemiskt namn eller material 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride)
Kvantitet 10 g
InChI formel InChI=1S/C9H11NO3.ClH/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;/h2-4,10-12H,5H2,1H3;1H
Formel vikt 217.0506