Läs mer
Adrenochrome Bisulfite Sodium Salt, TRC
Produktkod. 30324534
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30324534 10 mg
1 options
Produktkod. 30324534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA30525510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Adrenochrome Bisulfite Sodium Salt
CAS 5818-84-8
Molekylformel C9 H10 N O6 S . Na
Kvantitet 10 mg
Synonym 3aH-Indole-3a-sulfonic acid, 1,2,3,4,5,6-hexahydro-3-hydroxy-1-methyl-5,6-dioxo-, sodium salt (1:1), 3a(4H)-Indolinesulfonic acid, 5,6-dihydro-3-hydroxy-1-methyl-5,6-dioxo-, sodium salt (7CI), 3aH-Indole-3a-sulfonic acid, 1,2,3,4,5,6-hexahydro-3-hydroxy-1-methyl-5,6-dioxo-, monosodium salt (9CI), Adrenochrome bisulfite sodium salt, 1,2,3,4,5,6-Hexahydro-3-hydroxy-1-methyl-5,6-dioxo-3aH-indole-3a-sulfonic acid sodium salt
InChI formel InChI=1S/C9H11NO6S.Na/c1-10-4-8(13)9(17(14,15)16)3-6(12)5(11)2-7(9)10;/h2,8,13H,3-4H2,1H3,(H,14,15,16);/q;+1/p-1
LEDER [Na+].CN1CC(O)C2(CC(=O)C(=O)C=C12)S(=O)(=O)[O-]
IUPAC-namn sodium;3-hydroxy-1-methyl-5,6-dioxo-3,4-dihydro-2H-indole-3a-sulfonate
Molekylvikt (g/mol) 283.23
Formel vikt 283.0127