missing translation for 'onlineSavingsMsg'
Läs mer

Grant Instruments™ Advanced Aspirator with 2 L Trap Flask

The FTA-2i aspirator with 2L trap flask is designed for removal of alcohol, buffer and liquid from reaction vessels eg during DNA / RNA purification and other macromolecule re-precipitation techniques.

Brand:  Grant Instruments™ FTA-2I

Produktkod. 17191782

  • 15115.00 SEK / Paket med 1
Bara null kvar
Beräknad leverans: 27-04-2026
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

  • All in one system with integrated pump
  • 2 L autoclavable polypropylene trap flask
  • Fitted with hydrophobic microbiological filter
  • Vacuum pressure: -200 to -800 mbar (adjustable)
  • Overflow protection – liquid level sensor detects excess liquid and prevents overflow
  • The use of an aspirator is preferential to using a generic laboratory vacuum line as it restricts the possibility of contamination and can be used exactly where the operator is located.
  • Optional accessory : autoclavable hand operator for more comfortable usability of accessory adapters, featuring pressure sensitive button that controls aspiration speed.

Applications:

  • Aspiration/removal of alcohol/buffer from tube walls during DNA/RNA purification and other macromolecule reprecipitation techniques.
  • Routine operations of cells washing from culture medium and resuspension in buffer.
TRUSTED_SUSTAINABILITY
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.