Learn More
Advanced Instruments Osmometer Calibration Standards
Calibration standards for use with Advanced osmometers
Brand:  Advanced Instruments 3LA201
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15198606
  • 1264.20 SEK / Pack of 10
Sweden Stock
(immediate availability): 1
Add to basket
Specifications
Specifications
Osmometer Calibration Standard
Liquid
2000mOsm/kg
Room Temperature
Osmometer instrument calibration and quality control
Clear
Sodium Chloride 0.15% to 9.5%, 2-Hydroxybiphenyl Sodium Salt Tetrahydrate <0.1%
3D3, 3250 and 3320 osmometers
10 x 5mL