Learn More
OHAUS™ Adventurer™ Precision Balances
Adventurer™ Precision incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer™ is the most complete balance in its class.
13375.00 SEK - 17755.00 SEK
Specifications
Connection Type | RS232, USB Host, USB Device (included). GLP/GMP data output with real-time clock. |
---|---|
Display Type | Full-color VGA graphic touchscreen with user-controlled brightness |
Display Dimensions | 109 mm |
Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX) |
Stabilization Time | 1.500 sec. |
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Price | Quantity & Availability | ||||
15511936
|
OHAUS™
30206082 |
820 g | AX822/E |
16095.00 SEK
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15521936
|
OHAUS™
30206083 |
2200 g | AX2201 |
17755.00 SEK
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15531936
|
OHAUS™
30206084 |
2200 g | AX2201/E |
13375.00 SEK
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Fast stabilization and reliable operation are enhanced by the AutoCalTM option to ensure solid weighing performance and accurate, repeatable results for standard laboratory applications.
- The wide viewing angle color touchscreen provides access to all of Adventurer’s applications, while dual USB ports with GLP/GMP tracking capabilities assist in the monitoring and reporting of data.
- The 1 mg models have a two piece, top-mounted draftshield with doors that open wide, providing ample room for sample placement, while the 0.01 g and 0.1 g models feature large weighing pans.
Specifications
RS232, USB Host, USB Device (included). GLP/GMP data output with real-time clock. | |
109 mm | |
1.500 sec. | |
354 x 230 x 100 mm |
Full-color VGA graphic touchscreen with user-controlled brightness | |
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX) | |
Gram, Milligram, Carat, Ounce, Ounce Troy, Pound, Pennyweight, Grain, Newton, Momme, Mesghal, Hong Kong Tael, Singapore Tael, Taiwan Tael, Tical, Tola, Baht, Custom | |
175 mm |