missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Mouse anti-Human, Porcine, Clone: 7H2B11, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AEBP2 |
---|---|
Clone | 7H2B11 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877745
|
Proteintech
66141-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867745
|
Proteintech
66141-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody recognizes adult-specific larger form (52 kDa) and the embryo-specific smaller form (31 kDa).
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.Specifications
AEBP2 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AE binding protein 2, AEBP2, Zinc finger protein AEBP2 | |
AEBP2 | |
IgG1 | |
Protein G | |
Antibody |
7H2B11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Porcine | |
Q6ZN18 | |
100522805, 121536 | |
AEBP2 Fusion Protein Ag17905 | |
Primary | |
-20°C | |
AEBP2 |