Learn More
AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Packaging:
Poly Micro Tube
Description
This specific, irreversible inhibitor of serine proteases is a stable, nontoxic alternative to PMSF.
Specifications
Melting Point | 175°C |
Color | White |
Assay Percent Range | ≥98.0 % |
Packaging | Poly Micro Tube |
Quantity | 50 mg |
Formula Weight | 239.7 |
Percent Purity | ≥98 (HPLC) |
Physical Form | Powder or Solid |
Chemical Name or Material | AEBSF Hydrochloride |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.