missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFM Mouse anti-Human, Mouse, Clone: 2G9F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66113-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.Specifications
AFM | |
Monoclonal | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O89020, P43652 | |
Afm | |
AFM Fusion Protein Ag16866 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
2G9F6 | |
Unconjugated | |
AFM | |
afamin, afamin/AFM, AFM, ALB2, ALBA, ALF, Alpha Alb, Alpha albumin | |
Mouse | |
Protein G | |
RUO | |
173, 280662 | |
-20°C | |
Liquid |