missing translation for 'onlineSavingsMsg'
Learn More

Afuresertib, MedChemExpress

Product Code. 16409327
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Kvantitet:
5 mg
This item is not returnable. View return policy

Product Code. 16409327

missing translation for 'mfr': Medchem Express HY157275mg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

MedChemExpress Afuresertib (GSK2110183) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3, respectively.

Afuresertib (GSK2110183) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3, respectively.

Specifications

Kemiskt namn eller material Afuresertib
CAS 1047644-62-1
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C18H17Cl2FN4OS
Kvantitet 5 mg
Synonym GSK2110183
Löslighetsinformation DMSO : 100 mg/mL (234.02 mM; Need ultrasonic)
LEDER O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)S1)N[C@@H](CC3=CC=CC(F)=C3)CN
Molekylvikt (g/mol) 427.32
Formel vikt 427.32
Procent renhet 99.23%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.