Learn More
Xylem AG Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)
Product Code. 30484460
Change view
Click to view available options
Refractive Index:
1.33659
1.34401
1.34782
1.34968
1.35171
1.35568
Unit Size:
25mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index Unit Size
30484460 1.35171 25mL
30484478 1.33659 25mL
30484477 1.34401 25mL
30484445 1.34782 25mL
30484475 1.34968 25mL
30484466 1.35568 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30484460 Supplier Xylem Supplier No. 90507
  • ONLINE EXCLUSIVE PRICE
    2260.00 SEK
    1936.26 SEK / 25ml
    Save 323.74 SEK
    14% Av
View all promotions
 Restricted Item
I lager
Calibration fluid for calibration of refractometers.
  • Calibration Fluid
  • For Refractometers
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Fluid
Refractive Index 1.35171
For Use With (Equipment) Refractometer