missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agar, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 lb.
5 lb.
Description
- Light tan granular powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
Quantity | 1 lb. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'