missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16346-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The receptor for advanced glycation endproducts (RAGE) is a member of the Immunoglobin superfamily of cell surface markers. This protein is able to interact with many molecules including: advanced glycation endproducts (AGE), amphoterin, and ligands. Research has shown that the accumulation of RAGE ligands in the biological system (endothelium mononuclear phagocytes, neurons and smooth muscle cells) can lead to destructive tissue diseases such as diabetic retinopathy, amyloidoses, tumors and inflammation disorders.Specifications
AGER | |
Polyclonal | |
Unconjugated | |
AGER | |
AGER, RAGE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11596, 177, 81722 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15109, Q62151, Q63495 | |
AGER | |
AGER Fusion Protein Ag9540 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |