missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AGER |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825944
|
Proteintech
16346-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815944
|
Proteintech
16346-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The receptor for advanced glycation endproducts (RAGE) is a member of the Immunoglobin superfamily of cell surface markers. This protein is able to interact with many molecules including: advanced glycation endproducts (AGE), amphoterin, and ligands. Research has shown that the accumulation of RAGE ligands in the biological system (endothelium mononuclear phagocytes, neurons and smooth muscle cells) can lead to destructive tissue diseases such as diabetic retinopathy, amyloidoses, tumors and inflammation disorders.Specifications
AGER | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q15109, Q62151, Q63495 | |
11596, 177, 81722 | |
AGER Fusion Protein Ag9540 | |
Primary | |
-20°C | |
AGER |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AGER, RAGE | |
AGER | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |