Läs mer
AGI-6780, MedChemExpress
Produktkod. 16496778
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496778 10 mg
16479327 5 mg
16462857 50 mg
16416906 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496778 Leverantör Medchem Express Leverantörsnummer HY15734
MedChemExpress AGI-6780 that potently and selectively inhibits the tumor-associated mutant IDH2R140Q with IC50 of 23±1.7 nM. AGI-6780 is less potent against IDH2WT with IC50 of 190±8.1 nM.
AGI-6780 that potently and selectively inhibits the tumor-associated mutant IDH2R140Q with IC50 of 23±1.7 nM. AGI-6780 is less potent against IDH2WT with IC50 of 190±8.1 nM.
Specifikationer
Kemiskt namn eller material AGI-6780
CAS 1432660-47-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C21H18F3N3O3S2
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 29 mg/mL (60.23 mM)
LEDER O=S(C1=CC=C(C2=CSC=C2)C(NC(NC3=CC=CC(C(F)(F)F)=C3)=O)=C1)(NC4CC4)=O
Molekylvikt (g/mol) 481.51
Formel vikt 481.51
Procent renhet 97.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death