Läs mer
AGN 193109 Ethyl Ester, TRC
Produktkod. 30318517
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30318517 1 mg
1 options
Produktkod. 30318517 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA4270101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2-[5,6-Dihydro-5,5-dimethyl-8-(4-methylphenyl)-2-naphthalenyl]ethynyl]benzoic Acid Ethyl Ester
CAS 171568-43-7
Molekylformel C30H28O2
Kvantitet 1 mg
Synonym AGN 193109 Ethyl Ester, 4-[2-[5,6-Dihydro-5,5-dimethyl-8-(4-methylphenyl)-2-naphthalenyl]ethynyl]benzoic Acid Ethyl Ester
InChI formel InChI=1S/C30H28O2/c1-5-32-29(31)25-15-10-22(11-16-25)8-9-23-12-17-28-27(20-23)26(18-19-30(28,3)4)24-13-6-21(2)7-14-24/h6-7,10-18,20H,5,19H2,1-4H3
LEDER CCOC(=O)c1ccc(cc1)C#Cc2ccc3c(c2)C(=CCC3(C)C)c4ccc(C)cc4
IUPAC-namn ethyl 4-[2-[5,5-dimethyl-8-(4-methylphenyl)-6H-naphthalen-2-yl]ethynyl]benzoate
Molekylvikt (g/mol) 420.54