Läs mer
AGN 194310, MedChemExpress
Produktkod. 16471629
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
1mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16471629 1 mg 1mg
16463157 50 mg -
16433536 25 mg -
16429667 5 mg -
16427128 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16471629 Leverantör Medchem Express Leverantörsnummer HY16681
MedChemExpress AGN 194310 (VTP-194310) is a high affinity, potent and selective retinioic acid receptors (RARs) pan-antagonist with Kd values of 3 nM, 2 nM, 5 nM for RARα, RARβ, RARγ, respectively.
AGN 194310 (VTP-194310) is a high affinity, potent and selective retinioic acid receptors (RARs) pan-antagonist with Kd values of 3 nM, 2 nM, 5 nM for RARα, RARβ, RARγ, respectively.
Specifikationer
Kemiskt namn eller material AGN 194310
Fysisk form Solid
CAS 229961-45-9
Färg White
Kvantitet 1 mg
Anteckningar om renhetsgrad Research
Molekylformel C28H24O2S
För användning med (applikation) Cancer-programmed cell death
Synonym VTP-194310
Löslighetsinformation DMSO : 50 mg/mL (117.77 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER CC(S1)(C)C=C(C2=CC=C(CC)C=C2)C3=C1C=CC(C#CC4=CC=C(C(O)=O)C=C4)=C3
Molekylvikt (g/mol) 424.55
Formel vikt 424.55
Procent renhet 98.0%