missing translation for 'onlineSavingsMsg'
Learn More

AGO2 Mouse anti-Human, Clone: 4E2B2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66720-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828865

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

AGO2
4E2B2
Unconjugated
AGO2
Liquid
AGO2
AGO2 Fusion Protein Ag17723
Protein A
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UKV8
AGO2, Argonaute2, eIF 2C 2, eIF2C 2, EIF2C2, hAgo2, PAZ Piwi domain protein, PPD, Protein argonaute 2, Protein slicer, Q10
Mouse
IgG2b
20 μL
-20°C
Primary
27161
Videos
SDS
Documents
Certificates
Special Offers

Special Offers