missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGO2 Mouse anti-Human, Clone: 4E2B2, Proteintech
Mouse Monoclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | AGO2 |
---|---|
Clone | 4E2B2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828865
|
Proteintech
66720-1-IG-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16818865
|
Proteintech
66720-1-IG-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
AGO2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AGO2, Argonaute2, eIF 2C 2, eIF2C 2, EIF2C2, hAgo2, PAZ Piwi domain protein, PPD, Protein argonaute 2, Protein slicer, Q10 | |
AGO2 | |
IgG2b | |
Protein A | |
Antibody |
4E2B2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UKV8 | |
27161 | |
AGO2 Fusion Protein Ag17723 | |
Primary | |
-20°C | |
AGO2 |