missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10601-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein.Specifications
AGPAT1 | |
Polyclonal | |
Unconjugated | |
AGPAT1 | |
1 AGP acyltransferase 1, 1 AGPAT 1, 1 AGPAT1, AGPAT1, G15, LPAAT alpha, LPAATA, Protein G15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10554, 406165, 55979 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35083, Q99943 | |
Agpat1 | |
AGPAT1 Fusion Protein Ag0837 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |