missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR2 Mouse anti-Human, Porcine, Clone: 1A8A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66768-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).Specifications
AGR2 | |
Monoclonal | |
0.93 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95994 | |
AGR2 | |
AGR2 Fusion Protein Ag2919 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG2b |
Western Blot | |
1A8A8 | |
Unconjugated | |
AGR2 | |
AG 2, AG2, AGR2, GOB 4, HAG 2, HPC8, PDIA17, XAG 2 | |
Mouse | |
Protein A | |
RUO | |
100514482, 10551 | |
-20°C | |
Liquid |