Learn More
AHCY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10757-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.Specifications
AHCY | |
Polyclonal | |
Unconjugated | |
AHCY | |
Adenosylhomocysteinase, AdoHcyase, AHCY, S adenosylhomocysteine hydrolase, SAHH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
191, 269378, 29443 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10760, P23526, P50247 | |
AHCY | |
AHCY Fusion Protein Ag1208 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |