Läs mer
AICAR 3’,5’-Cyclic Phosphate, TRC
Produktkod. 30321187
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30321187 10 mg
30324610 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30321187 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA44050010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material AICAR 3',5'-Cyclic Phosphate
CAS 35908-14-6
Molekylformel C9 H13 N4 O7 P
Kvantitet 10 mg
InChI formel InChI=1S/C9H13N4O7P/c10-7-4(8(11)15)13(2-12-7)9-5(14)6-3(19-9)1-18-21(16,17)20-6/h2-3,5-6,9,14H,1,10H2,(H2,11,15)(H,16,17)/t3-,5-,6-,9-/m1/s1
LEDER NC(=O)c1c(N)ncn1[C@@H]2O[C@@H]3COP(=O)(O)O[C@H]3[C@H]2O
IUPAC-namn 3-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-5-aminoimidazole-4-carboxamide
Molekylvikt (g/mol) 320.2
Formel vikt 320.0522