missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIDA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23724-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
AIDA acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.Specifications
AIDA | |
Polyclonal | |
Unconjugated | |
AIDA | |
AIDA, C1orf80, RP11 378J18.7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108909, 64853 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C4Q6, Q96BJ3 | |
AIDA | |
AIDA Fusion Protein Ag20588 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |