missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ AIM2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142553
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
Specifikationer
| AIM2 | |
| Polyclonal | |
| Unconjugated | |
| AIM2 | |
| Absent in melanoma 2; AIM2; Gm1313; Ifi210; ifi-210; Interferon-inducible protein 210; interferon-inducible protein AIM2; Interferon-inducible protein p210; PYHIN4; RP11-520H16.3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O14862 | |
| AIM2 | |
| Peptide with sequence C-DKQYKSVTKPKPLSQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu