missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Aiolos/IKZF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NBP2-24495SS
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Aiolos/IKZF3 Polyclonal specifically detects Aiolos/IKZF3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifikationer
| Aiolos/IKZF3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 88% in dog, mouse and rat and 82% homologous in horse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 0.25 - 1.0 ug/mL, Immunohistochemistry reported in scientific literature (PMID 34207079), Immunocytochemistry/Immunofluorescence 1:100 | |
| Q9UKT9 | |
| IKZF3 | |
| A portion of amino acids 350-400 of human IKZF3 was used as the immunogen. | |
| 0.025 mg | |
| Chromatin Research | |
| 22806 | |
| Human, Primate | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering