Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Black/Yellow
Product Code. 17762068
Change view
Click to view available options
Dimensions (L x W):
91 x 60 cm
150 x 91 cm
1830 x 60 cm
1830 x 91 cm
1830 x 122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17762068 150 x 91 cm Each
17732068 91 x 60 cm Each
17742058 1830 x 60 cm Each
17772058 1830 x 91 cm Each
17702068 1830 x 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17762068 Supplier Notrax Supplier No. 410S0335BY
  • ONLINE EXCLUSIVE PRICE
    890.13 SEK
    723.91 SEK / Paket med 1
    Save 166.22 SEK
    18% Av
Beräknad leverans: 04-12-2026
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
  • Ribbed pattern enhances traction
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Dimensions (L x W) 150 x 91 cm
Color Black/Yellow
Length (English) 59 in.
Length (Metric) 150 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm