Läs mer
Ajmaline, TRC
Produktkod. 30319265
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30319265 100 mg
1 options
Produktkod. 30319265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA448000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ajmaline
CAS 4360-12-7
Molekylformel C20 H26 N2 O2
Kvantitet 100 mg
Synonym Ajmalan-17,21-diol, (17R,21α)-, Ajmaline (6CI,7CI,8CI), 5H-6,10:11,12a-Dimethanoindolo[3,2-b]quinolizine, ajmalan-17,21-diol deriv., (+)-Ajmaline, Ajmalin, Cardiorythmine, Gilurytmal, Ignazin, Merabitol, NSC 15627, Raugalline, Rauwolfin, Rauwolfine, Rhytmaton, Ritmos, Rytmalin, Siddiqui, Tachmalin, Takycor
InChI formel InChI=1S/C20H26N2O2/c1-3-10-11-8-14-17-20(12-6-4-5-7-13(12)21(17)2)9-15(16(11)18(20)23)22(14)19(10)24/h4-7,10-11,14-19,23-24H,3,8-9H2,1-2H3/t10-,11-,14-,15-,16+,17-,18+,19+,20+/m0/s1
LEDER CC[C@H]1[C@@H]2C[C@H]3[C@@H]4N(C)c5ccccc5[C@]46C[C@@H]([C@H]2[C@H]6O)N3[C@@H]1O
IUPAC-namn (1R,9R,10S,12R,13S,14R,16S,17S,18R)-13-ethyl-8-methyl-8,15-diazahexacyclo[14.2.1.01,9.02,7.010,15.012,17]nonadeca-2,4,6-triene-14,18-diol
Molekylvikt (g/mol) 326.43
Formel vikt 326.1994