missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKNA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23785-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.Specifications
AKNA | |
Polyclonal | |
Unconjugated | |
AKNA | |
AKNA, AT hook transcription factor, KIAA1968, RP11 82I1.4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80709 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z591 | |
AKNA | |
AKNA Fusion Protein Ag20662 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |