missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55230-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to AKT1. It has no cross reaction to AKT2 and AKT3.
The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.Specifications
AKT1 | |
Polyclonal | |
Unconjugated | |
AKT1 | |
AKT, AKT1, PKB, PKB ALPHA, PRKBA, Protein kinase B, Proto oncogene c Akt, RAC, RAC ALPHA, RAC PK alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11651, 207 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31749, P31750 | |
AKT1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |