missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ Akti-1/2

Description
Akti-1/2 is a potent and selective dual Akt1 and 2 inhibitor (IC50 values are 50 and 210 nM, respectively). Selective for Akt1 and 2 over a panel of other tyrosine and serine/threonine kinases. Sensitizes LnCaP cells to TRAIL (TNF-related apoptosis-inducing ligand) induced apoptosis. Also enhances CAR and TCR retroviral transduction of human T cells. Active in vivo.
Specifications
Specifications
| Kemiskt namn eller material | 1,3-Dihydro-1-[1-[[4-(6-phenyl-1 H-imidazo[4,5-g]quinoxalin-7-yl)phenyl]methyl]-4-piperidinyl]-2 H-benzimidazol-2-one |
| CAS | 612847-09-3 |
| Synonym | Akt Inhibitor VIII |
| Kvantitet | 50 mg |
| Mål | Akt (Protein Kinase B) Inhibitors |
| Molekylformel | C34H29N7O |
| Formel vikt | 551.64 |
| Renhet | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?