Läs mer
Al-23 Multibore Round Tube, O.D.: 5mm, Four 1mm Bores, Thermo Scientific™
Produktkod. 11342948
Kvantitet:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Förpackningsstorlek:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Produktkod. 11342948
Brand: Thermo Scientific Alfa Aesar 032710.DW
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Al-23 Multibore Round Tube
Namnnotering Four bores
Kvantitet 300 mm