Läs mer
Alatrofloxacin Mesylate, TRC
Produktkod. 30325586
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30325586 5 mg
1 options
Produktkod. 30325586 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5114005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alatrofloxacin Mesylate
CAS 146961-77-5
Molekylformel C26 H25 F3 N6 O5 . C H4 O3 S
Kvantitet 5 mg
Synonym Alatrofloxacin Mesylate, L-Alaninamide, L-alanyl-N-[(1α,5α,6α)-3-[6-carboxy-8-(2,4-difluorophenyl)-3-fluoro-5,8-dihydro-5-oxo-1,8-naphthyridin-2-yl]-3-azabicyclo[3.1.0]hex-6-yl]-, monomethanesulfonate (9CI), L-Alaninamide, L-alanyl-N-[3-[6-carboxy-8-(2,4-difluorophenyl)-3-fluoro-5,8-dihydro-5-oxo-1,8-naphthyridin-2-yl]-3-azabicyclo[3.1.0]hex-6-yl]-, (1α,5α,6α)-, monomethanesulfonate, 3-Azabicyclo[3.1.0]hexane, L-alaninamide deriv., Adrenochrome monoaminoguanidine mesilate, Alatrofloxacin mesylate, CP 116517, CP 116517-27, L-Alaninamide, L-alanyl-N-[(1α,5α,6α)-3-[6-carboxy-8-(2,4-difluorophenyl)-3-fluoro-5,8-dihydro-5-oxo-1,8-naphthyridin-2-yl]-3-azabicyclo[3.1.0]hex-6-yl]-, methanesulfonate (1:1)
InChI formel InChI=1S/C26H25F3N6O5.CH4O3S/c1-10(30)24(37)31-11(2)25(38)32-20-14-7-34(8-15(14)20)23-18(29)6-13-21(36)16(26(39)40)9-35(22(13)33-23)19-4-3-12(27)5-17(19)28;1-5(2,3)4/h3-6,9-11,14-15,20H,7-8,30H2,1-2H3,(H,31,37)(H,32,38)(H,39,40);1H3,(H,2,3,4)/t10-,11-,14-,15+,20+;/m0./s1
LEDER C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@H]1[C@@H]2CN(C[C@H]12)c3nc4N(C=C(C(=O)O)C(=O)c4cc3F)c5ccc(F)cc5F.CS(=O)(=O)O
IUPAC-namn 7-[(1R,5S)-6-[[(2S)-2-[[(2S)-2-aminopropanoyl]amino]propanoyl]amino]-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
Molekylvikt (g/mol) 654.61
Formel vikt 654.172