Läs mer
Albendazole-2-amino-sulphone-D3 Hydrochloride, TRC
Produktkod. 30179956
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179956 10 mg
30179933 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179956 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA51161410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1435902-07-0
Molekylformel C10 D3 H10 N3 O2 S . Cl H
Molekylvikt (g/mol) 278.774
Synonym Albendazole-2-aminosulfone D3 (Propyl-3,3,3 D3) Hydrochloride, 5-(3,3,3-Trideuteriopropylsulfonyl)-1H-benzimidazol-2-amine hydrochloride, Albendazole-2-aminosulfone D3 hydrochloride
IUPAC-namn 5-(3,3,3-trideuteriopropylsulfonyl)-1H-benzimidazol-2-amine;hydrochloride
LEDER Cl.[2H]C([2H])([2H])CCS(=O)(=O)c1ccc2[nH]c(N)nc2c1
Specifikationer
Kemiskt namn eller material Albendazole-2-amino-sulphone-D3 Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C10H13N3O2S.ClH/c1-2-5-16(14,15)7-3-4-8-9(6-7)13-10(11)12-8;/h3-4,6H,2,5H2,1H3,(H3,11,12,13);1H/i1D3;
Formel vikt 278.068