missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, No Preservative

For research use

Brand:  Alfa Aesar™ J65042.K2

UNSPSC : 12352200

Additional Details : CAS Number : 9048-46-8 Weight : 1.00000kg

 View more versions of this product

Product Code. 15476699

  • 13182.80 SEK / 1 L


Estimated Shipment date
from Supplier 31-08-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications

Specifications

35% w/v
Liquid
Bovine Serum
RUO
No
Liquid
1L
2° to 8°C
Albumin, Bovine, Fraction V, 35% soln., Reagent grade, Fatty Acid Free, no preservative
Documentation

Technical Documents