Learn More
Alfa Aesar™ Albumin, Bovine, Fraction V, 98%, Reagent Grade, pH 7.0
For research use
Brand:  Alfa Aesar™ J64100.30
Additional Details : CAS Number : 9048-46-8 Weight : 0.25000kg
 View more versions of this product
Product Code. 15496199
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Specifications
Specifications
Lyophilized Powder
7.0
250g
2° to 8°C
Albumin, Bovine, Fraction V, 98%, Reagent grade, pH 7.0
Lyophilized powder
Reagent grade, 98%
Bovine Serum
RUO
No
Videos
SDS
For Research Use Only