Learn More
Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low IgG, Chromatographically Purified
For research use
Brand:  Alfa Aesar™ J65470.09
UNSPSC : 12352200
Additional Details : CAS Number : 9048-46-8 Weight : 0.01000kg
 View more versions of this product
Product Code. 15476909
  • 242.20 SEK / 10g
Estimated Shipment date
from Supplier 31-08-2017
Add to basket
Disclaimers:
For Research Use Only
Specifications
Specifications
For immunoassays that require low Igg so as not to interfere with antibody-antigen binding
Lyophilized powder
Bovine Serum
RUO
No
Lyophilized Powder
10g
2° to 8°C
Albumin, Bovine, New Zealand origin, Low Igg, Chromatographically purified
Documentation
Technical Documents