missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Human, 96%

For research use

Brand:  Alfa Aesar™ J66780.ME

Additional Details : CAS Number : 70024-90-7 Weight : 0.00050kg

 View more versions of this product

Product Code. 15488269

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Specifications

Specifications

Produced in the liver, albumin is the most abundant protein in blood plasma. A carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Binds exogenous substances.
Lyophilized powder
500mg
2° to 8°C
Albumin, Human, 96%
Lyophilized Powder
96%
Human Serum
RUO
No
Videos
SDS

For Research Use Only